JWPH942F3BE2 J & W Pharmlab LLC

4-Methyl-pipecolinic acid

  • MDL number MFCD04118088
  • Cas Number : 172823-76-6
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1204

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1204
Availability details for
Quantity
  • Availability
  • Add to Cart