JWPH942F3BEF J & W Pharmlab LLC

(3S,5R)-5-Trifluoromethyl-piperidin-3-ol

  • MDL number MFCD31620789
  • Cas Number : 2165681-21-8
  • Molecular Weight : 169.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1367

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1367
Availability details for
Quantity
  • Availability
  • Add to Cart