JWPH942F3BFA J & W Pharmlab LLC

1-Benzyl-piperidine-4,4-dicarboxylic acid methyl ester

  • MDL number MFCD31812904
  • Cas Number : 2387534-94-1
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Light Yellow Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1388

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1388
Availability details for
Quantity
  • Availability
  • Add to Cart