JWPH942F3C48 J & W Pharmlab LLC

(3R,5R)-Piperidine-3,5-diol

  • MDL number MFCD22394876
  • Cas Number : 1043449-04-2
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1528

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1528
Availability details for
Quantity
  • Availability
  • Add to Cart