JWPH942F3C67 J & W Pharmlab LLC

1-(3-Trifluoromethoxy-phenyl)-piperazin-2-one

  • MDL number MFCD07373441
  • Cas Number : 215649-79-9
  • Molecular Weight : 260.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0107

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0107
Availability details for
Quantity
  • Availability
  • Add to Cart