JWPH942F3C80 J & W Pharmlab LLC

cis-1-Benzyl-3,5-dimethyl-piperazine

  • MDL number MFCD10566939
  • Cas Number : 55115-99-6
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0298

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0298
Availability details for
Quantity
  • Availability
  • Add to Cart