JWPH942F3C83 J & W Pharmlab LLC

3-Benzyl-1-methyl-piperazine

  • MDL number MFCD20427400
  • Cas Number : 137539-25-4
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0304

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0304
Availability details for
Quantity
  • Availability
  • Add to Cart