JWPH942F3C85 J & W Pharmlab LLC

1-Cyclohexyl-3-methyl-piperazine

  • MDL number MFCD00211067
  • Cas Number : 1368796-81-9
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0305

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0305
Availability details for
Quantity
  • Availability
  • Add to Cart