JWPH942F3CB2 J & W Pharmlab LLC

(1S,4S)-rel-2-Benzyl-2,5-Diaza-bicyclo[2.2.1]heptane

  • MDL number MFCD06411670
  • Cas Number : 1217977-97-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0332

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0332
Availability details for
Quantity
  • Availability
  • Add to Cart