JWPH942F3CC2 J & W Pharmlab LLC

1-Boc-4-(1-oxo-indan-5-yl)-piperazine

  • MDL number MFCD09701341
  • Cas Number : 954240-38-1
  • Molecular Weight : 316.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0404

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0404
Availability details for
Quantity
  • Availability
  • Add to Cart