JWPH942F3CC5 J & W Pharmlab LLC

(1S,4S)-2,5-Diazabicyclo(2.2.2)octane

  • MDL number MFCD09997868
  • Cas Number : 194600-16-3
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0424

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0424
Availability details for
Quantity
  • Availability
  • Add to Cart