JWPH942F3CC9 J & W Pharmlab LLC

6-Bromomethyl-pteridine-2,4-diamine

  • MDL number MFCD13180628
  • Cas Number : 59368-16-0
  • Molecular Weight : 255.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0486

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0486
Availability details for
Quantity
  • Availability
  • Add to Cart