JWPH942F3CE2 J & W Pharmlab LLC

3-(6,7-Dimethoxy-4-methyl-3-oxo-3,4-dihydro-quinoxalin-2-yl)-propionic acid

  • MDL number MFCD16293817
  • Cas Number : 132788-56-8
  • Molecular Weight : 292.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0484

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0484
Availability details for
Quantity
  • Availability
  • Add to Cart