JWPH942F3CE4 J & W Pharmlab LLC

Methyl-(6-methyl-imidazo[1,2-a]pyridin-2-ylmethyl)-amine dihydrochloride

  • MDL number MFCD03419478
  • Cas Number : 1187931-74-3
  • Molecular Weight : 248.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0187S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0187S
Availability details for
Quantity
  • Availability
  • Add to Cart