JWPH942F3CF2 J & W Pharmlab LLC

3-hydroxy-6-methoxyquinoxaline-2-carboxylic acid

  • MDL number MFCD22201142
  • Cas Number : 181529-97-5
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0501

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0501
Availability details for
Quantity
  • Availability
  • Add to Cart