JWPH942F3D03 J & W Pharmlab LLC

2-Phenyl-5-propyl-2H-pyrazol-3-ylamine

  • MDL number MFCD06738761
  • Cas Number : 1017781-36-0
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0134

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0134
Availability details for
Quantity
  • Availability
  • Add to Cart