JWPH942F3D07 J & W Pharmlab LLC

3,4-Dihydro-2H-quinoxaline-1,7-dicarboxylic acid 1-tert-butyl ester

  • MDL number MFCD11226331
  • Cas Number : 1185051-01-7
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0561

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0561
Availability details for
Quantity
  • Availability
  • Add to Cart