JWPH942F3D0A J & W Pharmlab LLC

[1,5]Diazocane-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11616045
  • Cas Number : 223797-64-6
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0564

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0564
Availability details for
Quantity
  • Availability
  • Add to Cart