JWPH942F3D12 J & W Pharmlab LLC

(R)-2-Trifluoromethyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD08686630
  • Cas Number : 1240588-01-5
  • Molecular Weight : 254.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form White Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0675

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0675
Availability details for
Quantity
  • Availability
  • Add to Cart