JWPH942F3D1B J & W Pharmlab LLC

2-Bromo-6-tert-butyl-pyrazine

  • MDL number MFCD10688561
  • Cas Number : 959239-49-7
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form 0
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0691

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0691
Availability details for
Quantity
  • Availability
  • Add to Cart