JWPH942F3D1E J & W Pharmlab LLC

6-Bromo-pyrazine-2-carbaldehyde

  • MDL number MFCD30011689
  • Cas Number : 1197227-82-9
  • Molecular Weight : 187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form 0
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0694

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0694
Availability details for
Quantity
  • Availability
  • Add to Cart