JWPH942F3D49 J & W Pharmlab LLC

3-Iodo-5-nitro-imidazo[1,2-a]pyridine

  • MDL number MFCD06796159
  • Cas Number : 885276-56-2
  • Molecular Weight : 289.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0281

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0281
Availability details for
Quantity
  • Availability
  • Add to Cart