JWPH942F3D6D J & W Pharmlab LLC

(8-Nitro-imidazo[1,2-a]pyridin-2-yl)-acetic acid

  • MDL number MFCD06739272
  • Cas Number : 59128-17-5
  • Molecular Weight : 221.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0290

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0290
Availability details for
Quantity
  • Availability
  • Add to Cart