JWPH942F3D78 J & W Pharmlab LLC

2H-Pyrazol-3-yl-methylamine

  • MDL number MFCD05667127
  • Cas Number : 37599-58-9
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0375

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0375
Availability details for
Quantity
  • Availability
  • Add to Cart