JWPH942F3D99 J & W Pharmlab LLC

4,6-Dichloro-cinnoline

  • MDL number MFCD27996597
  • Cas Number : 68211-13-2
  • Molecular Weight : 199.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0615

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0615
Availability details for
Quantity
  • Availability
  • Add to Cart