JWPH942F3DFD J & W Pharmlab LLC

5-Ethyl-1-phenyl-1H-pyridin-2-one

  • MDL number MFCD09997893
  • Cas Number : 53427-98-8
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0340

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0340
Availability details for
Quantity
  • Availability
  • Add to Cart