JWPH942F3E01 J & W Pharmlab LLC

2-Cyclopentyloxy-nicotinonitrile

  • MDL number MFCD09946128
  • Cas Number : 1016805-97-2
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0368

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0368
Availability details for
Quantity
  • Availability
  • Add to Cart