JWPH942F3E02 J & W Pharmlab LLC

2-Cyclohexyloxy-nicotinic acid

  • MDL number MFCD09742983
  • Cas Number : 68359-02-4
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0370

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0370
Availability details for
Quantity
  • Availability
  • Add to Cart