JWPH942F3E2D J & W Pharmlab LLC

2-Bromo-6-chloro-4-trifluoromethyl-pyridine

  • MDL number MFCD18255893
  • Cas Number : 823221-94-9
  • Molecular Weight : 260.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0881

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0881
Availability details for
Quantity
  • Availability
  • Add to Cart