JWPH942F3E8E J & W Pharmlab LLC

2-Iodo-4,6-dimethyl-nicotinonitrile

  • MDL number MFCD22689334
  • Cas Number : 1163297-83-3
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0972

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0972
Availability details for
Quantity
  • Availability
  • Add to Cart