JWPH942F3EA1 J & W Pharmlab LLC

4-Hydroxy-pyridine-2,6-dicarboxylic acid dimethyl ester

  • MDL number MFCD00185834
  • Cas Number : 19872-91-4
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0923

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0923
Availability details for
Quantity
  • Availability
  • Add to Cart