JWPH942F3EA9 J & W Pharmlab LLC

2-Bromo-4-ethyl-5-nitro-pyridine

  • MDL number MFCD12024433
  • Cas Number : 929617-28-7
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R0947

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R0947
Availability details for
Quantity
  • Availability
  • Add to Cart