JWPH942F3EBD J & W Pharmlab LLC

Furo[2,3-c]pyridine-2-carbaldehyde

  • MDL number MFCD15143904
  • Cas Number : 112372-06-2
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1023

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1023
Availability details for
Quantity
  • Availability
  • Add to Cart