JWPH942F3EE7 J & W Pharmlab LLC

2-Pyrrolidin-2-yl-pyrimidin-4-ylamine

  • MDL number MFCD18432635
  • Cas Number : 1263378-09-1
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/70R0212

Safety Information

MSDS https://www.jwpharmlab.com/home/product/70R0212
Availability details for
Quantity
  • Availability
  • Add to Cart