JWPH942F3EF5 J & W Pharmlab LLC

(R)-4-Amino-4-pyridin-4-yl-butyric acid

  • MDL number MFCD11035366
  • Cas Number : 1213334-54-3
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1421

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1421
Availability details for
Quantity
  • Availability
  • Add to Cart