JWPH942F3F16 J & W Pharmlab LLC

6-Methyl-5-trifluoromethyl-pyridin-2-ylamine

  • MDL number MFCD09881222
  • Cas Number : 1023813-33-3
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1268

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1268
Availability details for
Quantity
  • Availability
  • Add to Cart