JWPH942F3F70 J & W Pharmlab LLC

3-Chloro-6-pyridin-2-yl-pyridazine

  • MDL number MFCD12143147
  • Cas Number : 78784-70-0
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/70R0279

Safety Information

MSDS https://www.jwpharmlab.com/home/product/70R0279
Availability details for
Quantity
  • Availability
  • Add to Cart