JWPH942F3F96 J & W Pharmlab LLC

5,6-Dimethyl-7-(1-phenyl-ethyl)-3,7-dihydro-pyrrolo[2,3-d]pyrimidin-4-one

  • MDL number MFCD12196882
  • Cas Number : 82703-45-5
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/70R0415

Safety Information

MSDS https://www.jwpharmlab.com/home/product/70R0415
Availability details for
Quantity
  • Availability
  • Add to Cart