JWPH942F3FA0 J & W Pharmlab LLC

2-Oxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester

  • MDL number MFCD10688281
  • Cas Number : 33458-27-4
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/70R0429

Safety Information

MSDS https://www.jwpharmlab.com/home/product/70R0429
Availability details for
Quantity
  • Availability
  • Add to Cart