JWPH942F3FB1 J & W Pharmlab LLC

3-Ethoxy-pyrrolidine

  • MDL number MFCD08687759
  • Cas Number : 45592-49-2
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0218

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0218
Availability details for
Quantity
  • Availability
  • Add to Cart