JWPH942F3FBE J & W Pharmlab LLC

3-Pyrimidin-5-yl-benzylamine

  • MDL number MFCD11932683
  • Cas Number : 1181409-07-3
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/70R0466

Safety Information

MSDS https://www.jwpharmlab.com/home/product/70R0466
Availability details for
Quantity
  • Availability
  • Add to Cart