JWPH942F3FE7 J & W Pharmlab LLC

Ethyl 4,5-diphenyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD04115125
  • Cas Number : 73799-68-5
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/72R0007

Safety Information

MSDS https://www.jwpharmlab.com/home/product/72R0007
Availability details for
Quantity
  • Availability
  • Add to Cart