JWPH942F3FE8 J & W Pharmlab LLC

(4,5,6,7-Tetrahydro-1H-pyrrolo[3,2-c]pyridin-4-yl)-acetic acid ethyl ester

  • MDL number MFCD11506294
  • Cas Number : 82859-02-7
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/72R0015

Safety Information

MSDS https://www.jwpharmlab.com/home/product/72R0015
Availability details for
Quantity
  • Availability
  • Add to Cart