JWPH942F4006 J & W Pharmlab LLC

(2S, 4S)-1-N-Boc-4-bromo-proline benzyl ester

  • MDL number MFCD09701397
  • Cas Number : 874162-95-5
  • Molecular Weight : 384.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0169

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0169
Availability details for
Quantity
  • Availability
  • Add to Cart