JWPH942F4009 J & W Pharmlab LLC

(1R,3R,4S)-1-Aza-bicyclo[2.2.1]hept-3-ylamine

  • MDL number MFCD11858559
  • Cas Number : 508209-68-5
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0265

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0265
Availability details for
Quantity
  • Availability
  • Add to Cart