JWPH942F401D J & W Pharmlab LLC

(S)-3-(Pyrrolidin-3-yloxy)-phenylamine

  • MDL number MFCD10567058
  • Cas Number : 853213-27-1
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0222

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0222
Availability details for
Quantity
  • Availability
  • Add to Cart