JWPH942F402C J & W Pharmlab LLC

2-Aza-spiro[4.4]nonane-2,4-dicarboxylic acid 2-benzyl ester

  • MDL number MFCD20278357
  • Cas Number : 1823295-96-0
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0417

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0417
Availability details for
Quantity
  • Availability
  • Add to Cart