JWPH942F403B J & W Pharmlab LLC

(4S)-2-Aza-spiro[4.5]decane-4-carboxylic acid

  • MDL number MFCD20656262
  • Cas Number : 788143-23-7
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0446

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0446
Availability details for
Quantity
  • Availability
  • Add to Cart