JWPH942F4040 J & W Pharmlab LLC

(2S,4S)-4-Amino-2-methoxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD19441504
  • Cas Number : 348165-54-8
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0578

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0578
Availability details for
Quantity
  • Availability
  • Add to Cart