JWPH942F4069 J & W Pharmlab LLC

6-Iodo-quinolin-4-ylamine

  • MDL number MFCD06658311
  • Cas Number : 40107-08-2
  • Molecular Weight : 270.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/80R0074

Safety Information

MSDS https://www.jwpharmlab.com/home/product/80R0074
Availability details for
Quantity
  • Availability
  • Add to Cart